Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer pH 2.00 ± 0.01 at 25°C Reference Standard, Reagents

Catalog No. 01337442
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-442 4 L
01-337-445 10 L
01-337-444 1 L
01-337-443 20 L
01-337-441 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-337-442 Supplier Reagents Holdings Llc Supplier No. C21554004A
Add to Cart
Add to Cart

Buffer pH 2.00 ± 0.01 at 25°C Reference Standard, Reference Buffer, use for pH meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Buffer pH 2.00 ± 0.01 at 25°C Reference Standard
CAS 7732-18-5, 7447-40-7, 7647-01-0
Melting Point 0°C
Boiling Point 100°C
Color Colorless
For Use With (Application) Calibration
For Use With (Equipment) pH Meter
pH 2
Physical Form Liquid
Quantity 4 L
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 1.001
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.