Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer pH 7.00 ± 0.01 at 25°C, Color-Coded Yellow Reference Standard, Reagents

Catalog No. 01337415
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-337-415 100 mL Poly Bottle
01-337-414 10 L Cubitainer
01-337-413 1 L Poly Bottle
01-337-411 20 L Cubitainer
01-337-410 4 L Poly Bottle
01-337-409 4 L Cubitainer
01-337-408 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01-337-415 Supplier Reagents Holdings Llc Supplier No. C2156000100S
Add to Cart
Add to Cart

Buffer pH 7.00 ± 0.01 at 25°C, Color-Coded Yellow Reference Standard, Reference Buffer, use for pH meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Buffer pH 7.00 ± 0.01 at 25°C, Color-Coded Yellow Reference Standard
CAS 7732-18-5, 7558-79-4, 7778-77-0, 26628-22-8
Melting Point 0°C
Boiling Point 100°C
Color Yellow
For Use With (Application) Calibration
For Use With (Equipment) pH Meter
pH 7
Physical Form Liquid
Quantity 100 mL
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 1.006
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.