Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer pH 7, Green, Reagents

Catalog No. 01335733
Change view
Click to view available options
Quantity:
500 mL
4 L
10 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-733 20 L
01-335-734 10 L
01-335-732 4 L
01-335-731 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-335-733 Supplier Reagents Holdings Llc Supplier No. CB16750020F
Add to Cart
Add to Cart

Buffer pH 7, Green, Reagent, use for pH Meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Buffer pH 7, Green
CAS 7732-18-5, 7558-79-4, 7558-79-4
Boiling Point 100°C
Color Green
For Use With (Application) Calibration
For Use With (Equipment) pH Meter
pH 7
Physical Form Liquid
Quantity 20 L
Solution Type Buffer
Packaging Cubitainer
Specific Gravity 1.006
Sterility Non-sterile
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.