Learn More
Buffer, Reference Standard, pH 10.00 ± 0.01 at 25°C (Color Coded Blue), Ricca Chemical

Shop All Ricca Chemical Company Products


Specifications
Specifications
| Chemical Name or Material | Buffer, Reference Standard |
| Name Note | pH 10.00 ± 0.01 at 25 deg C (Color Coded Blue) |
| Melting Point | 0°C |
| CAS | 497-19-8, 144-55-8, 7732-18-5 |
| Boiling Point | 100°C |
| CAS Min % | 97.0, 0.51, 0.39 |
| Color | Blue |
| CAS Max % | 100.0, 0.53, 0.41 |
| Concentration | 1X |
| For Use With (Application) | Calibration |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.