Learn More
Buffer, Reference Standard, pH 11.00 ± 0.01 at 25°C, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Chemical Name or Material | Ricca Chemical Company Buffer pH 11.00 |
| CAS Min % | 97.0, 1.02, 0.11 |
| CAS Max % | 100.0, 1.06, 0.11 |
| CAS | 7558-79-4, 1310-73-2 |
| Concentration | 1X |
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 11 |
| Physical Form | Liquid |
| Quantity | 20 L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.