Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer, Reference Standard, pH 12.00 ± 0.01 at 25°C, Ricca Chemical
SDP

Catalog No. R161500010F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
10 L
20 L
Packaging:
Clear PET-G Bottle
Cubitainer
Natural Poly Bottle
Pack
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R161500010F 10 L Cubitainer
R161500020P 20 x 20 mL Pack
1615-16 500 mL Clear PET-G Bottle
161532 1 L Natural Poly Bottle
16151 4 L Natural Poly Bottle
16155 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. R161500010F Supplier Ricca Chemical Company Supplier No. R161500010F
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 12.00± 0.01 at 25°C
Melting Point 0°C
CAS 7447-40-7, 1310-73-2, 7732-18-5
Boiling Point 100°C
CAS Min % 98.0, 0.36, 0.08
Color Colorless
CAS Max % 100.0, 0.38, 0.08
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 12
Physical Form Liquid
Quantity 10 L
Solution Type pH Buffer
Packaging Cubitainer
Sterility Non-sterile
Solubility Information Miscible
Grade Reference Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.