Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer, Reference Standard, pH 6.00 ± 0.01 at 25°C, Ricca Chemical
BIOPHARMASDP

Catalog No. 151016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Pack
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
1510-16 500 mL Natural Poly Bottle
R151000020 20 x 20 mL Pack
1510-32 1 L Natural Poly Bottle
15101CT 4 L Cubitainer
1510-1 4 L Natural Poly Bottle
1510-25 10 L Cubitainer
R15105 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 1510-16 Supplier Ricca Chemical Company Supplier No. 151016
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 6.00± 0.01 at 25°C
CAS 7778-77-0, 7558-79-4, 7732-18-5
CAS Min % 97.0, 0.78, 0.1
Color Colorless
CAS Max % 100.0, 0.82, 0.11
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 6
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Packaging Natural Poly Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.