Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution pH 10.0 (20°C), Violet colored, borax / sodium hydroxide, Solstice

Catalog No. 60046297 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
10 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-297 6 x 1 L
60-024-80 500 mL
60-024-79 1 L
60-046-298 6 x 500 mL
60-024-78 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-046-297 Supplier Solstice Supplier No. 336686X1L
Add to Cart
Add to Cart

Violet colored, borax / sodium hydroxide

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 100°C
CAS Min % ≥90.0000%, ≥0.1000%
CAS Max % ≤100.0000%, <1.0000%
CAS 1330-43-4, 1310-73-2
Quantity 6 x 1 L
Solution Type Buffer
Packaging Poly Bottle
Chemical Name or Material Buffer solution pH 10.0 (20°C), Violet colored, borax / sodium hydroxide
ShelfLife 1260 days from date of manufacture
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.