Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution, 12.0, di-sodium hydrogen phosphate / sodium hydroxide solution, Solstice

Catalog No. 60048362 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
6 x 100 mL
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-362 6 x 1 L
60-024-64 100 mL
60-048-361 6 x 100 mL
60-024-65 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-048-362 Supplier Solstice Supplier No. 336516X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

di-Sodium hydrogen phosphate / sodium hydroxide solution

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name azanium;hydroxide
SMILES N.O

Specifications

Density 1.00g/mL at 20°C
For Use With (Application) pH Calibration
pH 12
Physical Form Liquid
Quantity 6 x 1 L
Solution Type pH Buffer
UN Number NONH for all modes of transport
Grade Not Available
Chemical Name or Material Buffer solution, di-sodium hydrogen phosphate / sodium hydroxide solution
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.