Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution pH 9.0 (20°C), Buffer solution pH 9.0 (20°C), Solstice

Catalog No. 60046291 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
10 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-291 6 x 1 L
60-024-57 500 mL
60-024-56 1 L
60-024-55 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-046-291 Supplier Solstice Supplier No. 336486X1L
Add to Cart
Add to Cart

Borax / hydrochloric acid, traceable to SRM from NIST

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Synonym Sodium tetraborate buffer solution

Specifications

CAS Max % ≤100.0000%, <1.0000%
CAS Min % ≥0.1000%
Density 1 g/cm3
Quantity 6 x 1 L
Solution Type Buffer
Packaging Poly Bottle
MDL Number MFCD00081185
Chemical Name or Material Buffer solution pH 9.0 (20°C), Buffer solution pH 9.0 (20 °C)
ShelfLife 1260 days from date of manufacture
Recommended Storage Room Temp
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.