Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution B, for low level Sulfate Analysis (Turbidimetric Method), Ricca Chemical
SDP

Catalog No. 14768 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14768 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14768 Supplier Ricca Chemical Company Supplier No. 14768
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Analysis of Low-Level Sulfate (Turbidimetric Method)

Specifications

Color Colorless
Physical Form Liquid
Quantity 250 mL
Packaging Natural Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.