Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Solution II - Buffer Solution, for Microbiology (Eosinophil Staining)

Catalog No. ES6922
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6922 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES6922 Supplier ENG Scientific Supplier No. 6922
Add to Cart
Add to Cart

Used in staining eosinophils from secretions of the nasal passage, sinuses, bronchi and urinary sediment.

  • For Eosinophil Stain Kit
  • Contains Solution I - Eosin Y, Solution II - Buffer Solution, and Solution III - Methylene Blue Solution

Specifications

CAS 1336-21-6
Quantity 250 mL
Molecular Formula H5NO
Packaging Bottle
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name amine hydrate
Molecular Weight (g/mol) 35.046

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.