Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution, pH 1.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable

Catalog No. AA40446AP
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40446AP 500 mL
AA40446K2 1 L
AA40446K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40446AP Supplier Thermo Scientific Chemicals Supplier No. 040446AP
Add to Cart

For Calibration of pH Meters | CAS: 7447-40-7

Calibration of pH meters

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution
CAS 7447-40-7, 7647-01-0, 7732-18-5
Color Colorless
Density 1.005
Physical Form Liquid
Quantity 500 mL
MDL Number MFCD00146252
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Concentration or Composition (by Analyte or Components) Potassium chloride: 0.7%; Hydrochloric acid: 0.7%; water: 98.6%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.