Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution pH 11.0 at 25°C, Certified, LabChem™

Catalog No. LC125402
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC125402 1 L
LC125401 500 mL
LC125404 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC125402 Supplier LabChem, Inc. Supplier No. LC125402
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Buffer Solution pH 11.0 at 25°C
Color Colorless
CAS Max % 99.64, 0.35, 0.01
CAS 7558-79-4, 1310-73-2
Decomposition Information Phosphine; Phosphorus oxides; Phosgene
Density 1g/mL
pH 11.00 ±0.05
Physical Form Liquid
Quantity 1 L
Identification Passes Test
Linear Formula Na2HPO4
Packaging Poly Bottle
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 141.96
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.