Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution, pH 11.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable

Catalog No. AA42424AP
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42424AP 500 mL
AA42424K2 1 L
AA42424K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42424AP Supplier Thermo Scientific Chemicals Supplier No. 042424AP
Add to Cart

CAS: 1336-21-6 | H5NO | 35.05 g/mol

Buffer solution (pH 11) is used as a means of keeping pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution
CAS 1336-21-6
Color Colorless
Physical Form Liquid
Quantity 500 mL
Molecular Formula H5NO
Odor Odorless
MDL Number MFCD00066650
Solubility Information Miscible with water.
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name amine hydrate
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.