Learn More
Buffer Solution pH 13.0 at 25°C, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Buffer Solution pH 13.0 at 25°C |
| Color | Brown to Colorless |
| CAS Max % | 99.24, 0.37, 0.34, 0.05 |
| CAS | 7447-40-7, 1310-73-2, 89-83-8 |
| Decomposition Information | Phosphine; Phosphorus oxides; Phosgene; Thermal decomposition generates: Corrosive vapors; Carbon dioxide; Carbon monoxide |
| Density | 1g/mL |
| pH | 13.00 ±0.05 |
| Physical Form | Liquid |
| Quantity | 4 L |
| Identification | Passes Test |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.