Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution Ph 2.0 (20 °C), Solstice

Catalog No. 6000387 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
6 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-003-87 1 L
60-003-86 100 mL
60-048-364 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-003-87 Supplier Solstice Supplier No. 310451L
Add to Cart
Add to Cart

Citric acid / hydrochloric acid / sodium chloride, with fungicide, traceable to SRM from NIST and PTB

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer Solution Ph 2.0 (20 °C)
Name Note With fungicide
CAS 7732-18-5, 7647-14-5, 77-92-9
Quantity 1 L
Packaging Plastic Bottle
MDL Number MFCD00146206
UN Number 1789
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.