Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution, pH 4.010 (+/-0.022 @ 25°C), Colored Red, Specpure, NIST Traceable

Catalog No. AA38713M4
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38713M4 20 L
AA38713AP 500 mL
AA38713K2 1 L
AA38713K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA38713M4 Supplier Thermo Scientific Chemicals Supplier No. 038713M4
Add to Cart

CAS: 1310-73-2

Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications. Buffer solutions are essential to maintain the suitable condition to keep the correct pH for enzymes in many organisms to work. It is also used in fermentation processes and in setting the correct conditions for dyes used in coloring fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution
CAS 1310-73-2, 7778-77-0, 7732-18-5
Color Red
Physical Form Liquid
Quantity 20 L
MDL Number MFCD00146206
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Solubility Information Miscible with water.
Concentration or Composition (by Analyte or Components) Sodium hydroxide: 0.7%; Potassium dihydrogen phosphate: 0.7%; water: 98.6%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.