Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 4.00, Color-Coded Pink, Mettler Toledo™
GSA_VA

Catalog No. 01917117 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-917-117 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-917-117 Supplier METTLER TOLEDO Supplier No. 63056121
Add to Cart
Add to Cart

METTLER TOLEDO™ solutions for electrode calibration and care.

  • Quality solutions for accurate results
  • Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions
  • Each standard solution comes with a certificate of analysis, providing traceability to NIST standards

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 0°C
CAS 877-24-7
Boiling Point 100°C
Color Red
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 4.01
Physical Form Liquid
Quantity 500 mL
Solution Type Calibration
Molecular Formula C8H5KO4
Synonym Potassium Biphthalate
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
IUPAC Name potassium;2-carboxybenzoate
Molecular Weight (g/mol) 204.222
PubChem CID 23676735
Formula Weight 204.23
Chemical Name or Material Buffer Solution, pH 4.00
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.