Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution pH 6.86 at 25°C, Certified, LabChem™

Catalog No. LC123502
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer™
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC123502 1 L Poly Bottle
LC123501 500 mL Poly Bottle
LC123504 4 L Poly Bottle
LC123506 10 L Cubitainer™
LC123505 20 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC123502 Supplier LabChem, Inc. Supplier No. LC123502
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Buffer Solution pH 6.86 at 25°C
Color Colorless
CAS Max % 99.25, 0.68, 0.07
CAS 7778-77-0, 1310-73-2
Decomposition Information Phosphorus oxides; Sodium oxide
pH 6.86 ±0.01
Physical Form Liquid
Quantity 1 L
Identification Passes Test
Linear Formula KH2PO4
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 136.09
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.