Learn More
Buffer Solution pH 7.0 at 25°C, Color Coded Yellow, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Buffer Solution pH 7.0 at 25°C |
| Name Note | Color Coded Yellow |
| Color | Yellow |
| CAS Max % | 99.24, 0.68, 0.08 |
| CAS | 7778-77-0, 1310-73-2 |
| Decomposition Information | Phosphorus oxides; Sodium oxide |
| pH | 7.00 ±0.01 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Identification | Passes Test |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.