Learn More
Buffer solution pH 7.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable
Description
Used to determine pH of solutions in various industrial applications. Buffer solutions of known and accurate pH value allows to adjust testing system to measure the samples with unknown pH more precisely and accurately.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Buffer solution |
| CAS | 1336-21-6 |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Molecular Formula | H5NO |
| Odor | Odorless |
| MDL Number | MFCD00066650 |
| Traceability to NIST | Yes |
| Solubility Information | pH 7.00 buffers are in solution form. Miscible with water. |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.