Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, pH 7.00 ± 0.01 at 25°C, Reference Standard, Spectrum™ Chemical
SDP

Catalog No. 18609342 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-342 500 mL Poly Bottle
18-609-343 1 L Poly Bottle
18-609-344 4 L Poly Bottle
18-609-345 20 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-609-342 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B1165500MLPL
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Buffer Solution, pH 7.00 +/- 0.01 @ 25 DEG C, Reference Standard
CAS 7778-77-0, 1310-73-2, 7732-18-5
For Use With (Application) pH Measurement
For Use With (Equipment) pH Meter
pH 7.00 ±0.01
Physical Form Solution
Quantity 500 mL
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.