Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Buffer, pH 7, for Residual Chlorine and Chlorine Dioxide (Amperometric), Ricca Chemical
SDP

Catalog No. 580316 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5803-16 500 mL
5803-1 4 L
5803-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 5803-16 Supplier Ricca Chemical Company Supplier No. 580316
Add to Cart
Add to Cart

For APHA Testing

Specifications

Chemical Name or Material Phosphate Buffer
CAS 7732-18-5, 7558-79-4, 7778-77-0
For Use With (Application) APHA for Chlorine and Chlorine Dioxide (Amperometric Method)
pH 7
Physical Form Solution
Quantity 500 mL
Packaging Natural Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.