Learn More
G-Biosciences STM BUFFER (270MM SUCROSE, 10MM TRIS.HCL, PH 7.5, 1MM MGCL2), 1L
Supplier:  G-Biosciences 786537
Catalog No. 50-103-5440
Only null left
Add to Cart
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.