Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Buffers for RNA Transctription and Post-Transcriptional Modifications

Catalog No. p-200022102
Change view
Click to view available options
Quantity:
200 μL
800 μL
1000 μL
Chemical Name or Material:
Capping Buffer
Poly(A) Polymerase Buffer
T4 RNA Ligase 2 Reaction Buffer
T4 RNA Ligase Reaction Buffer
Transcription Buffer
Concentration:
10X
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Chemical Name or Material Concentration
FERF8156AF 200 μL Transcription Buffer 10X
FERB23AF 800 μL Capping Buffer 10X
FERB2348AF 1000 μL Poly(A) Polymerase Buffer 10X
FERB1299AF 200 μL T4 RNA Ligase Reaction Buffer 10X
FERB3277AF 200 μL T4 RNA Ligase 2 Reaction Buffer 10X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. FERF8156AF Supplier Thermo Scientific™ Supplier No. F8156AF
Only null left
Add to Cart
Add to Cart

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Features

  • Convenient stand-alone enzymes and buffers for workflow optimization
  • No animal-origin components in final product composition
  • Reliable and reproducible results with rigorous quality testing
  • Scalable and available as OEM products

Applications

  • CRISPR/Cas-based genome editing
  • Gene expression
  • Generation of pluripotent cells
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Transcription Buffer
Concentration 10X
For Use With (Application) RNA Synthesis
pH 8
Physical Form Liquid
Quantity 200 μL
Includes 10X Transcription Buffer, 200 μL

Store at -20°C.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.