Learn More
Selleck Chemical LLC Buflomedil HCl S2607-50mg
SDP
Supplier:  Selleck Chemical LLC S260750MG
Encompass_Preferred
Buflomedil HCl(NSC 759291) is a vasodilator used to treat claudication or the symptoms of peripheral arterial disease A nonselective alpha adrenergic receptor inhibitor
Catalog No. 50-311-2370
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.