Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BugBuster™ 10X Protein Extraction Reagent

Catalog No. 709213 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10mL
50mL
100mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-921-3 10mL
70-921-4 50mL
70-921-5 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 70-921-3 Supplier MilliporeSigma™ Supplier No. 709213
Add to Cart
Add to Cart

Reagent for protein sample preparation

  • Concentrated formulation of proprietary detergents without addition of salts or buffer components
  • Provides flexible alternative to ready-to-use standard 1X BugBuster, allowing user-defined dilution and addition of buffer components
  • Consists all of the bioprocessing benefits of standard BugBuster plus the freedom to control pH, reagent concentration, and buffer additives necessary for maximum extraction and activity of your target protein
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10mL
Content And Storage Store at+15°C to+30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.