Learn More
Description
- Combines BugBuster Protein Extraction Reagent and Benzonase™ Nuclease in one convenient reagent
- Soluble proteins are gently extracted from E. coli without exposure to heat or oxidative damage
- Viscosity is eliminated by nucleic acid digestion in a single step
- The resulting protein extract can easily be fractionated by conventional purification techniques
- Ideally suited for application in high throughput protein purifications
Specifications
Specifications
| Quantity | 500mL |
| Content And Storage | Store at +15° to +30°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.