Learn More
Description
- The BugBuster and rLysozyme efficiently lyse the bacterial cells while the Benzonase degrades nucleic acids to facilitate downstream processing
- Use 5mL BugBuster and 10μL Lysonase per 1g cell paste
- The kits provide sufficient reagents for protein extraction from 20g and 100g cell paste
Specifications
Specifications
| Quantity | 1 kit |
| Kit Contents | BugBuster Protein Extraction Reagent (100mL), Lysonase Bioprocessing Reagent (200μL) |
| Content And Storage | Multiple storage requirements |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.