Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BugBuster™ Protein Extraction Reagent

Catalog No. 705841000ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-584-1000ML 1000 mL
70-584-4 500 mL
70-584-3 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 70-584-1000ML Supplier MilliporeSigma™ Supplier No. 705841000ML
Add to Cart
Add to Cart

Reagent for protein sample preparation

  • Formulated for gentle disruption of cell wall of E. coli, resulting in liberation of soluble protein
  • Provides simple, rapid, low cost alternative to mechanical methods
  • Formulation utilizes mixture of non-ionic detergents capable of cell wall perforation without denaturing soluble protein
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1000 mL
Content And Storage Store at 15°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.