Learn More
S-Curve™ BD-25 Bulk Dispenser with 2 Compartments – Extra Large
Catalog No. 19162417 Shop All S-Curve Technologies Products
Catalog No. 19-162-417 Supplier S-Curve™ Supplier No. BD25
Add to Cart
Add to Cart
Multiple use opportunities for gloves, face masks, hair nets, bouffants, shoe covers, etc.
  • Available in 1/4 in. clear acrylic or 1/4 in. clear PETG material – safe with IPA
  • Open top for easy access to product
  • Flame treated edges for smoothness
  • Rubber pads on bottom corners to prevent slipping & scratching
Specifications
Product Type Bulk Dispenser
For Use With (Equipment) For use with gloves, face masks, hair nets, bouffants, shoe covers