Learn More
Poltex Bulk Bin, 4 in. Wide Comp
SDP
Catalog No. 21377522
Change view
Click to view available options
Mounting Type:
Magnetic Mount
Tape Mount
Wall Mount or Countertop
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
21-377-522 Wall Mount or Countertop
21-377-520 Magnetic Mount
21-377-521 Tape Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-377-522 Supplier Poltex Supplier No. BLKBN4W
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Simple 4 in. bulk bin with open top for filling and front opening for dispensing.
Simple Bulk Bin with open top for filling and front for dispensing.
Durable and cleanable PETG construction.
Choose from:
  • Wall/countertop mount with rubber bumpons on base
  • VHB tape mount (very high bond tape included)
  • Magnet mount (powerful magnets included)
Specifications
Type Dispensing Bin
Material PETG
Color Clear
Shape Rectangular
Mounting Type Wall Mount or Countertop
Autoclavable Not Autoclavable