Learn More
Poltex Bulk Bin With Lid, 8 in. Wide Comp
SDP
Catalog No. 21377535
Change view
Click to view available options
Mounting Type:
Magnetic Mount
Tape Mount
Wall Mount or Countertop
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
21-377-535 Magnetic Mount
21-377-536 Tape Mount
21-377-537 Wall Mount or Countertop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-377-535 Supplier Poltex Supplier No. BLKBNLD8M
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Simple 8 in. bulk bin with lid, open top with lid for filling, and front opening for dispensing.
Simple bulk bin with lid
Durable and cleanable PETG construction
8 in. wide
Choose from:
  • Wall or countertop mount with rubber bumpons on base
  • VHB tape mount (very high bond tape included)
  • Magnet mount (powerful magnets included)
Specifications
Type Dispensing Bin
Material PETG
Color Clear
Shape Square
Mounting Type Magnetic Mount
Autoclavable Not Autoclavable