Learn More
Poltex Bulk Cone Mask Dispenser

Description
Clear dispenser holds a box or stack of cone masks. Poltex Blue Cone Mask Dispenser is has a hinged lid to keep dust out.
- Holds a stack of masks or the whole box.
- Made from durable 3/16 in. thick PETG
- Hinged lid keeps dust out
- Interior DxWxH: 5.5 x 5.5 x 7 in.
- Clear
- Available with magnets or holes for mounting
Specifications
Specifications
| Color | Clear |
| Material | PETG |
| For Use With (Application) | Storing and dispensing cone masks |
| Dimensions (D x W x H) Interior | 5.5 x 5.5 x 7 in. |
| Mounting | Pre-drilled Holes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.