Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Bulk Conical Tube Bin
SDP

Catalog No. 22000394
Change view
Click to view available options
Mounting:
Keyhole slots for wall mount
Powerful Magnet Mount
Powerful VHB Tape Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-000-394 Powerful Magnet Mount
22-000-395 Powerful VHB Tape Mount
22-000-396 Keyhole slots for wall mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-000-394 Supplier Poltex Supplier No. BULKMOUNTM

Bulk tube bin to help organize hood and/or bench

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Bulk Conical Tubes, Test Tubes, Microcentrifuge Tubes
Material PETG
Positions 1
Sterility Non-sterile
For Use With (Equipment) Bulk conical tubes, test tubes, microcentrifuge tubes
Mounting Powerful Magnet Mount
Quantity 1
Length (Metric) 205 mm
Weight (Metric) 250 g
Includes Bin
Height (Metric) 221 mm
Width (Metric) 185 mm
Show More Show Less

Consumables not included.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.