Learn More
Poltex Bulk Earloop Mask Disp Mount
Description
Holds a stack of bulk earloop masks. Slanted internal ramp helps to feed masks toward the front. Choose from magnet mount, VHB tape mount, or wall mount.
- Durable, cleanable PETG construction
- VHB tape mount (very high bond tape included)
- Wall mount (screws and anchors included)
- Magnet mount (powerful magnets included)
Specifications
Specifications
| Product Type | Face Mask Dispenser |
| Includes | Screws and Anchors for Mounting |