Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Bulk Micropipette Tips
SCI_ED

Catalog No. S39854 Shop All Eisco Products
Change view
Click to view available options
Volume (Metric):
0.2 to 10 μL
10,000 μL
2.0 to 200 μL
200 to 1000 μL
5000 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S39854 0.2 to 10 μL
S39855 2.0 to 200 μL
S39856 200 to 1000 μL
S39857 5000 μL
S39858 10,000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S39854 Supplier Eisco™ Supplier No. CH0587A
Add to Cart
Edge
Add to Cart

Eisco™ Bulk Micropipette Tips are designed to be used with variable or fixed single and multi channel micropipettes.

  • Autoclavable
  • Free of lubricants, dyes and heavy metals
  • Non-sterile
  • Supplied in resealable plastic bag

Specifications

Volume (Metric) 0.2 to 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (English) 1.3 in.
Length (Metric) 3.3 cm
For Use With (Application) Electrophoresis, PCR
Format Bulk
Tip Style Fine Point
Packaging Bag
Quantity 1000
Product Type Filter Tips
Disposable Single-use
For Use With (Equipment) Universal Fit Micro
Graduated No
Material Plastic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.