Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco Labs Bulk Pack of Narrow Mouth HDPE Amber Reagent Bottles with Polypropylene Caps
SCI_ED

Catalog No. S000110 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000110 500 mL
S000109 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000110 Supplier Eisco Labs Supplier No. CH5438PK125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bulk pack of narrow-mouth amber HDPE reagent bottles with polypropylene caps are UV-protective, leak-tested, and chemically resistant, ideal for storing light-sensitive liquids in lab or industrial environments.

  • Bulk pack: Narrow-mouth amber HDPE reagent bottles with polypropylene caps, designed for secure storage and transport of sensitive liquids and samples.
  • Light shielding design: Amber color protects light-sensitive samples and reagents from UV exposure, making these bottles ideal for lab and industrial applications.
  • Excellent chemical resistance: HDPE material withstands exposure to various chemicals, ensuring reliable use in laboratory and industrial settings.
  • Leak-proof tested: Rigorously tested caps and bottle threads ensure no leaks, providing safe and secure storage for liquids and reagents.
  • Lightweight and durable: Made from lightweight, durable HDPE, these translucent amber bottles allow for visibility of contents while providing protection and ease of use. Non-sterile and not autoclavable. Protects light-sensitive substances with a temperature range of -100 to +120°C.

Specifications

Capacity (Metric) 500 mL
Color Amber
Diameter (Metric) Inner Neck 21.6 mm
Material HDPE
No. per Pack 125 Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 170 mm
Diameter (Metric) 73.5 mm
Autoclavable Not Autoclavable
Capacity (English) 17 oz.
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.