Learn More
Eisco Labs Bulk Pack of Narrow Mouth HDPE Translucent Reagent Bottles with Polypropylene Caps
Shop All Eisco Products

Description
- Bulk pack of reagent bottles - Includes narrow mouth translucent HDPE bottles with polypropylene caps. Ideal for large-scale lab or industrial applications.
- Chemical resistant bottles - Constructed from HDPE for excellent resistance to a variety of chemicals, making these bottles versatile for storing reagents, solutions, and samples.
- Leak-proof tested for reliability - Each bottle is rigorously tested to ensure no leaks under pressure. Perfect for safe storage and transport of liquids in laboratory and industrial settings.
- Lab-friendly features - Narrow mouth for controlled pouring. Translucent design allows easy monitoring of liquid levels during use.
- Suited for extreme conditions - Durable and reliable with a temperature range of -100°C to +120°C. Perfect for diverse applications in laboratories, research facilities, and industrial environments.
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Capacity (English) | 17 oz. |
| Color | Translucent |
| Diameter (Metric) Inner Neck | 21.6 mm |
| Material | HDPE |
| No. per Pack | 125 Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Narrow |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.