Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco Labs Bulk Pack of Wide Mouth HDPE Reagent Bottles with Polypropylene Caps
SCI_ED

Catalog No. S000140 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000140 500 mL
S000139 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000140 Supplier Eisco Labs Supplier No. CH5535PK125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bulk pack of wide-mouth HDPE reagent bottles with polypropylene caps are chemically resistant, leak-tested, and easy to fill and handle, making them ideal for high-volume lab, industrial, or sample storage applications.

  • Bulk pack: Includes bulk pack of wide-mouth HDPE reagent bottles with polypropylene caps, designed for safe liquid storage and lab applications.
  • Designed for versatility: Wide-mouth design makes filling and pouring liquids easy, ensuring convenience for lab, industrial, or sample storage applications.
  • Chemical resistance: HDPE construction offers excellent resistance to a variety of chemicals, making these bottles suitable for diverse laboratory uses. Designed for secure liquid storage with a -100 to 120°C temperature range.
  • Leak-proof tested: Ensures safe transport and storage with rigorously tested caps and bottle threads to prevent leaks, even under pressure.
  • Lightweight and translucent: Lightweight HDPE material allows for easy handling, while the translucent body offers visibility of contents.

Specifications

Capacity (Metric) 500 mL
Color Translucent
Diameter (Metric) Inner Neck 43.7 mm
Material HDPE
No. per Pack 125 Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide
Height (Metric) 167 mm
Diameter (Metric) 73.7 mm
Autoclavable Not Autoclavable
Capacity (English) 17 oz.
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.