Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Bulk PPE Dispenser - Bump, Wall Mount
SDP

Catalog No. 01259160
Change view
Click to view available options
Dimensions (L x W x H):
12 x 18 x 16.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
01-259-160 12 x 18 x 16.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
N/A
Catalog No. 01-259-160 Supplier Poltex Supplier No. BULKPPE181217W
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
N/A

Holds and dispenses bulk PPE with top fill and lower front dispensing. Wall mount design with durable and cleanable PETG construction.

  • Holds and dispenses bulk PPE
  • Top-fill design with lower front dispensing
  • Wall mount (screws and anchors included)
  • Durable and cleanable PETG construction
  • Consumables not included

Specifications

Product Type PPE Dispenser
Color Clear
Dimensions (L x W x H) 12 x 18 x 16.5 in.
Holds Bouffant Caps, Shoe Covers, PPE, Isolation Gowns or Coveralls, PPE
Material PETG
Mounting Wall Mount
No. of Compartments 1
Product Style Hinged Lid with Front Opening
Edges Rounded
For Use With (Application) Holds, Dispenses Bulk PPE
Includes Wall Mount Hardware
Quantity 1 Ea.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.