Learn More
Wanzl Bulk Shelving Boxed Steel Frame
Description
- Boxed Steel Frame
- Replacement part for bulk shelving system
Specifications
Specifications
| Includes | 1 Boxed Steel Frame |
| Height (English) | 60 in. |
| Color | Chrome |
| Depth (English) | 36 in. |
| For Use With (Application) | Shelving |
| Material | Steel |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.