Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Bulk tubes
GREENER_CHOICE

Catalog No. 15340161 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Non-reinforced Tubes
Reinforced Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
15-340-161 Non-reinforced Tubes
15-340-162 Reinforced Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-340-161
Add to Cart
Add to Cart

For use with Bead Mill 4 and Bead Mill 24 homogenizers. Also fits most other commercially available Bead Mills.

  • RNase/DNase free
  • Processing range: 300μL to 1.8mL
  • Beads not included

Specifications

Product Type Non-reinforced Tubes
Capacity (Metric) 2 mL
Color White/Opaque
Dnase Free Yes
For Use With (Equipment) Bead Mill 4 and 24 Homogenizers
Includes Screw Caps and O-rings
Packaging Quantity 500/Pk.
RNase Free Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.