Learn More
Bullard™ ADP20 Air Driven Pump
SDP
Catalog No. 189991825 Shop All Bullard Company Products
Change view
Click to view available options
For Use With (Equipment):
4 hoods or 6 half or full facemasks
Catalog No. For Use With (Equipment)
18-999-1825 4 hoods or 6 half or full facemasks
1 options
Catalog No. 18-999-1825 Supplier Bullard™ Supplier No. ADP20
  • $5,756.00
    $4,825.65 / Each of 1
    Save $930.35
    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request a Quote
Continuous flow, non-electric air pump does not require CO monitors, alarms, or calibration procedures.
  • ADP20 is ideal for users working in locations without an electrical power source
  • For use in potentially explosive work environments
  • Provides 30 cfm of air operating at 5 psig
Specifications
For Use With (Equipment) 4 hoods or 6 half or full facemasks
Includes Pressure gauge, two 0.5 in. industrial interchange (Hansen compatible) Q.D. couplers
Model ADP20
Weight (English) 200 lb.
Product Type Breathing Air Pump