Learn More
Bullard™ SAR Air Filter CO Monitor 5 PPM
SDP
Catalog No. 19824414 Shop All Bullard Company Products
Change view
Click to view available options
Includes:
2-outlet unit
6-outlet unit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-824-414 2-outlet unit
19-824-412 6-outlet unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-824-414 Supplier Bullard™ Supplier No. CO5HP41P2
SAR filter with Carbon Monoxide (CO) monitoring available for use with 41 series 2-outlet or 6-outlet systems.
  • 41 Series Portable Filtration and CO Monitoring Kits
  • 5 PPM
  • 75 CFM
  • Available for 2-outlet or 6-outlet systems
Specifications
Alarm Types Audio, Visual
Certifications/Compliance OSHA
Connection Type Hansen outlet couplings
Flow Rate 75 cfm
For Use With (Equipment) 41 Series Airline Respirator
Filter Type Heavy-duty 41 series filter with 7-stage filtration
Includes 2-outlet unit
Model CO5HP41P2
Pressure 5 ppm
Electrical Requirements 110 V
Product Type CO Monitor and Filter Kit