Learn More
Bullard™ Replacement Facepiece Assemblies for Spectrum Series Airline Respirators
SDP
Catalog No. 19807911 Shop All Bullard Company Products
Change view
Click to view available options
For Use With (Equipment):
Spectrum pressure demand respirator
Spectrum series continuous flow airline respirator
Size:
Medium/Large
Small
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Size
19-807-911 Spectrum pressure demand respirator Medium/Large
19-824-325 Spectrum series continuous flow airline respirator Small
19-807-909 Spectrum series continuous flow airline respirator Medium/Large
19-822-181 Spectrum pressure demand respirator Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-807-911 Supplier Bullard™ Supplier No. SPECPDML
This item is not returnable. View return policy
Used for respiratory protection
Specifications
Product Type Facepiece Assembly
Size Medium/Large
For Use With (Application) Painting, chemical handling, pharmaceuticals, confined space
For Use With (Equipment) Spectrum pressure demand respirator
Includes Facepiece, lens, head straps, exhalation valve, facepiece components
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.