Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ Portable Clean Air Boxes (CAB) Breathing Air Filtration Systems, One Worker Outlet
SDP

Catalog No. 19100606 Shop All Bullard Company Products
Change view
Click to view available options
Connection Type:
CEJN Outlet Couplings
Schrader Outlet Couplings
Snap-Tite Outlet Couplings
Stainless Steel Hansen Outlet Couplings
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
19-100-606 Stainless Steel Hansen Outlet Couplings
19-100-604 CEJN Outlet Couplings
19-100-605 Snap-Tite Outlet Couplings
19-100-603 Schrader Outlet Couplings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-100-606 Supplier Bullard™ Supplier No. CAB15SS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Portable clean air boxes offer 3-stage filtration with carbon monoxide (CO) monitoring to comply with OSHA’s requirement for providing workers with Grade D breathing air.

  • Clean Air Boxes (CAB) portable units made of polycarbonate
  • 15 cfm with one worker outlet made of a variety of materials
  • Offer 3-stage filtration with carbon monoxide (CO) monitoring
  • Provide workers with Grade D breathing air
  • Three stages of filtration remove oil, water, particulate, oil vapor and odors
  • Designed for multiple users with or without independent regulators in a portable or fixed option

Specifications

Color Yellow
Connection Type Stainless Steel Hansen Outlet Couplings
Flow Rate 15 cfm
For Use With (Equipment) CAB 15 cfm Breathing Air Filtration System
Holds 1 worker
Includes Pelican case, 3-stage filtration, visual filter change indicators, CO monitor with auto calibration, audible and visual alarms for high CO and low battery, and 6 ft. 110 V power cord
Model CAB15SS
Product Type Portable Clean Air Box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.