Learn More
Bullard™ Clean Air Box (CAB) Series Fixed Breathing Air Filtration/CO Monitoring, One to Eight Worker System
SDP
Catalog No. 19100669 Shop All Bullard Company Products
Change view
Click to view available options
Product Type:
Independent Regulator
Manifold Assembly
Connection Type:
CEJN outlet couplings
Hansen outlet couplings
Schrader outlet couplings
Snap-Tite™ outlet couplings
Stainless Steel Hansen Outlet Couplings
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Connection Type
19-100-669 Independent Regulator Hansen outlet couplings
19-100-664 Manifold Assembly Hansen outlet couplings
19-100-672 Independent Regulator Snap-Tite™ outlet couplings
19-100-667 Manifold Assembly Snap-Tite™ outlet couplings
19-100-673 Independent Regulator Stainless Steel Hansen Outlet Couplings
19-100-668 Manifold Assembly Stainless Steel Hansen Outlet Couplings
19-100-670 Independent Regulator Schrader outlet couplings
19-100-665 Manifold Assembly Schrader outlet couplings
19-100-671 Independent Regulator CEJN outlet couplings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
These units will not remove Carbon Monoxide (CO), Carbon Dioxide (CO2), Nitrogen, or other toxic gases or fumes. These units will not increase the oxygen content of an air supply and should not be used when air entering the system is oxygen deficient.
Catalog No. 19-100-669 Supplier Bullard™ Supplier No. CAB100PMIRHA
Add to Cart
Add to Cart
Request a Quote
These units will not remove Carbon Monoxide (CO), Carbon Dioxide (CO2), Nitrogen, or other toxic gases or fumes. These units will not increase the oxygen content of an air supply and should not be used when air entering the system is oxygen deficient.
Wall-mount clean air boxes offer 3-stage filtration with carbon monoxide (CO) monitoring to comply with OSHA’s requirement for providing workers with Grade D breathing air.
  • Clean Air Boxes (CAB) wall units made of stainless steel
  • 100 cfm with eight outlets
  • Offer 3-stage filtration with carbon monoxide (CO) monitoring
  • Provide workers with Grade D breathing air
  • Three stages of filtration remove oil, water, particulate, oil vapor and odors
  • Designed for multiple users with or without independent regulators in a portable or fixed option
Specifications
Color Yellow
Connection Type Hansen outlet couplings
Flow Rate 100 cfm
Material Stainless Steel
Includes 110 V Power Cord
Model CAB100PMIRHA
Product Type Independent Regulator