Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ Carbon Monoxide (CO) Monitoring System
SDP

Catalog No. 19100675 Shop All Bullard Company Products
Change view
Click to view available options
Model:
COHP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
19-100-675 COHP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-100-675 Supplier Bullard™ Supplier No. COHP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monitor analyzes the breathing air and displays the CO concentration.

  • Monitoring system
  • Displays green led light for normal operation and red led light for high CO levels
  • Includes lights, connection kit, and alarm
  • Uses 9V battery or 110 VAC power cord

Specifications

Model COHP
Pressure 10 ppm
Product Type CO Monitor System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.